Product Details

SNP ID
rs61730325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34759070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAATCTTTCGATGGTGAGTGAA[C/T]GATGAACTGTGGCTGAAGGTCTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF599 PubMed Links

Gene Details

Gene
ZNF599
Gene Name
zinc finger protein 599
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007248.2 2119 Silent Mutation TCA,TCG S577S NP_001007249.1
XM_017026369.1 2119 Silent Mutation TCA,TCG S540S XP_016881858.1

View Full Product Details