Product Details

SNP ID
rs34121431
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:91410201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGCTTTGTGGAGAGTGTTTAAT[C/G]ACAGAAATACAACTACTTGACACCA
Phenotype
MIM: 605327
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
NFIL3 PubMed Links

Gene Details

Gene
NFIL3
Gene Name
nuclear factor, interleukin 3 regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289999.1 1067 Silent Mutation GTC,GTG V178V NP_001276928.1
NM_001290000.1 1067 Silent Mutation GTC,GTG V178V NP_001276929.1
NM_005384.2 1067 Silent Mutation GTC,GTG V178V NP_005375.2
XM_017014743.1 1067 Silent Mutation GTC,GTG V178V XP_016870232.1
XM_017014744.1 1067 Silent Mutation GTC,GTG V178V XP_016870233.1

View Full Product Details