Product Details

SNP ID
rs6503857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58271636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCACAGCCACCCAGCGGCCCAC[A/G]ACGCCTGCCCCTCCTCACCGATCAC
Phenotype
MIM: 150205 MIM: 606989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LPO PubMed Links

Gene Details

Gene
LPO
Gene Name
lactoperoxidase
There are no transcripts associated with this gene.

Gene
MPO
Gene Name
myeloperoxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000250.1 Intron NP_000241.1

View Full Product Details