Product Details

SNP ID
rs34336938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21198346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCATACGGCATAGAGTTCGTAA[G/A]TCTTCTTTACACAGTACACCAGGGC
Phenotype
MIM: 616160
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DHRS7B PubMed Links

Gene Details

Gene
DHRS7B
Gene Name
dehydrogenase/reductase 7B
There are no transcripts associated with this gene.

Gene
TMEM11
Gene Name
transmembrane protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003876.2 1000 Missense Mutation ACT,ATT T186I NP_003867.1

View Full Product Details