Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_025104.3 | Intron | NP_079380.2 | |||
NM_145663.2 | Intron | NP_663696.1 | |||
XM_006722098.2 | Intron | XP_006722161.1 | |||
XM_006722102.3 | Intron | XP_006722165.1 | |||
XM_011525281.2 | Intron | XP_011523583.1 | |||
XM_017025145.1 | Intron | XP_016880634.1 | |||
XM_017025146.1 | Intron | XP_016880635.1 | |||
XM_017025147.1 | Intron | XP_016880636.1 | |||
XM_017025148.1 | Intron | XP_016880637.1 | |||
XM_017025149.1 | Intron | XP_016880638.1 | |||
XM_017025150.1 | Intron | XP_016880639.1 | |||
XM_017025151.1 | Intron | XP_016880640.1 | |||
XM_017025152.1 | Intron | XP_016880641.1 | |||
XM_017025153.1 | Intron | XP_016880642.1 | |||
XM_017025154.1 | Intron | XP_016880643.1 | |||
XM_017025155.1 | Intron | XP_016880644.1 | |||
XM_017025156.1 | Intron | XP_016880645.1 |