Product Details
- SNP ID
-
rs62636658
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67821204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGCTGGACCTGGGCACAGATTC[C/T]ACCCAGGAACTAAGGCTGCAGGTCA
- Phenotype
-
MIM: 611510
MIM: 607720
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CENPT
PubMed Links
Gene Details
- Gene
- CENPT
- Gene Name
- centromere protein T
There are no transcripts associated with this gene.
- Gene
- TSNAXIP1
- Gene Name
- translin associated factor X interacting protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001288990.1 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
NP_001275919.1 |
NM_001288991.1 |
295 |
Missense Mutation |
CAC,TAC |
H85Y |
NP_001275920.1 |
NM_001288992.1 |
295 |
Intron |
|
|
NP_001275921.1 |
NM_001288993.1 |
295 |
UTR 5 |
|
|
NP_001275922.1 |
NM_001288994.1 |
295 |
Intron |
|
|
NP_001275923.1 |
NM_018430.3 |
295 |
Silent Mutation |
TCC,TCT |
S68S |
NP_060900.2 |
XM_005256049.4 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_005256106.1 |
XM_011523230.2 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521532.1 |
XM_011523231.2 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521533.1 |
XM_011523232.2 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521534.1 |
XM_011523233.2 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521535.1 |
XM_011523234.2 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521536.1 |
XM_011523235.1 |
295 |
Silent Mutation |
TCC,TCT |
S68S |
XP_011521537.1 |
XM_011523236.2 |
295 |
Missense Mutation |
CAC,TAC |
H85Y |
XP_011521538.1 |
XM_011523237.2 |
295 |
Intron |
|
|
XP_011521539.1 |
XM_011523238.1 |
295 |
Intron |
|
|
XP_011521540.1 |
XM_011523239.1 |
295 |
Silent Mutation |
TCC,TCT |
S122S |
XP_011521541.1 |
XM_011523240.1 |
295 |
UTR 5 |
|
|
XP_011521542.1 |
XM_011523241.1 |
295 |
UTR 5 |
|
|
XP_011521543.1 |
XM_011523242.1 |
295 |
Intron |
|
|
XP_011521544.1 |
XM_011523243.1 |
295 |
UTR 5 |
|
|
XP_011521545.1 |
XM_017023445.1 |
295 |
Missense Mutation |
CAC,TAC |
H85Y |
XP_016878934.1 |
XM_017023446.1 |
295 |
Intron |
|
|
XP_016878935.1 |
XM_017023447.1 |
295 |
Missense Mutation |
CAC,TAC |
H85Y |
XP_016878936.1 |
XM_017023448.1 |
295 |
Intron |
|
|
XP_016878937.1 |
XM_017023449.1 |
295 |
Missense Mutation |
CAC,TAC |
H85Y |
XP_016878938.1 |
XM_017023450.1 |
295 |
UTR 5 |
|
|
XP_016878939.1 |
XM_017023451.1 |
295 |
UTR 5 |
|
|
XP_016878940.1 |
XM_017023452.1 |
295 |
Intron |
|
|
XP_016878941.1 |
XM_017023453.1 |
295 |
UTR 5 |
|
|
XP_016878942.1 |
XM_017023454.1 |
295 |
Intron |
|
|
XP_016878943.1 |
XM_017023455.1 |
295 |
Intron |
|
|
XP_016878944.1 |
View Full Product Details