Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001012267.2 | 1184 | Intron | NP_001012267.1 | ||
NM_001286969.1 | 1184 | Intron | NP_001273898.1 | ||
NM_001286971.1 | 1184 | Intron | NP_001273900.1 | ||
XM_011518685.2 | 1184 | Intron | XP_011516987.1 | ||
XM_011518689.1 | 1184 | Intron | XP_011516991.1 | ||
XM_017014715.1 | 1184 | Intron | XP_016870204.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014057.4 | 1184 | UTR 3 | NP_054776.1 | ||
NM_024416.4 | 1184 | UTR 3 | NP_077727.3 | ||
NM_033014.3 | 1184 | UTR 3 | NP_148935.1 |