Product Details

SNP ID
rs28730833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34255475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCACATTGTCTTCTCTCTGAGT[A/T]ATCATGGACCATCCAAGCAGACGTG
Phenotype
MIM: 158105
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCL2 PubMed Links

Gene Details

Gene
CCL2
Gene Name
C-C motif chemokine ligand 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002982.3 Intron NP_002973.1

View Full Product Details