Product Details

SNP ID
rs34163698
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133255883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGACCATCATGGCCTGGTGGCAG[A/G]CCCTGGTGAGCCGCTGCACCTCTTG
Phenotype
MIM: 110300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABO PubMed Links

Gene Details

Gene
ABO
Gene Name
ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020469.2 873 Missense Mutation GCC,GTC A283V NP_065202.2

View Full Product Details