Product Details

SNP ID
rs62637641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35751379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTAGCTGAGCTGGCTGGGGAA[C/T]GCCTAATGGCTGCCACATCCTGCCG
Phenotype
MIM: 609471 MIM: 612191 MIM: 108961 MIM: 615742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBA2 PubMed Links

Gene Details

Gene
GBA2
Gene Name
glucosylceramidase beta 2
There are no transcripts associated with this gene.

Gene
MSMP
Gene Name
microseminoprotein, prostate associated
There are no transcripts associated with this gene.

Gene
NPR2
Gene Name
natriuretic peptide receptor 2
There are no transcripts associated with this gene.

Gene
RGP1
Gene Name
RGP1 homolog, RAB6A GEF complex partner 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080496.2 749 Missense Mutation CGC,TGC R201C NP_001073965.2

View Full Product Details