Product Details

SNP ID
rs113618401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38707880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATCGCTTCAACAAGGTCAGCG[A/G]CCCCCCGCCGTGTCCCCTACCAGTT
Phenotype
MIM: 600328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105371763 PubMed Links

Gene Details

Gene
LOC105371763
Gene Name
uncharacterized LOC105371763
There are no transcripts associated with this gene.

Gene
MIR4734
Gene Name
microRNA 4734
There are no transcripts associated with this gene.

Gene
MLLT6
Gene Name
MLLT6, PHD finger domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005937.3 Intron NP_005928.2
XM_006721911.2 Intron XP_006721974.1
XM_017024648.1 Intron XP_016880137.1
XM_017024649.1 Intron XP_016880138.1
XM_017024650.1 Intron XP_016880139.1

View Full Product Details