Product Details

SNP ID
rs73051950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:25600945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTGCTATGCCTCCTTTGGACTA[C/T]ATTCAGCACTGATTCCTAGACACAC
Phenotype
MIM: 180220 MIM: 126431
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RARB PubMed Links

Gene Details

Gene
RARB
Gene Name
retinoic acid receptor beta
There are no transcripts associated with this gene.

Gene
TOP2B
Gene Name
topoisomerase (DNA) II beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001068.3 Intron NP_001059.2
XM_005265427.3 Intron XP_005265484.1
XM_011534057.2 Intron XP_011532359.1

View Full Product Details