Product Details

SNP ID
rs35200098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133100311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAGCAGCTCATAGTCCTCCGG[C/T]TCCTCCTCCTCCAGGTTGTGTTTGT
Phenotype
MIM: 601619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RALGDS PubMed Links

Gene Details

Gene
RALGDS
Gene Name
ral guanine nucleotide dissociation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042368.2 2606 Silent Mutation GAA,GAG E787E NP_001035827.1
NM_001271774.1 2606 Silent Mutation GAA,GAG E813E NP_001258703.1
NM_001271775.1 2606 Silent Mutation GAA,GAG E841E NP_001258704.1
NM_001271776.1 2606 Silent Mutation GAA,GAG E830E NP_001258705.1
NM_006266.3 2606 Silent Mutation GAA,GAG E842E NP_006257.1

View Full Product Details