Product Details

SNP ID
rs62638734
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:100720593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCCCCATTTTTCTCTTGGAGAA[C/T]CCAGATAGATAGTCCACTGAATGGG
Phenotype
MIM: 192225
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VCAM1 PubMed Links

Gene Details

Gene
VCAM1
Gene Name
vascular cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001078.3 403 Missense Mutation ACC,ATC T61I NP_001069.1
NM_001199834.1 403 Intron NP_001186763.1
NM_080682.2 403 Missense Mutation ACC,ATC T61I NP_542413.1

View Full Product Details