Product Details

SNP ID
rs145783420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11437811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGGGTCAGGGGCTCTTATCTGT[A/G]GATGGATGGGACATGTCTGTCCCTG
Phenotype
MIM: 615956 MIM: 177060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC151 PubMed Links

Gene Details

Gene
CCDC151
Gene Name
coiled-coil domain containing 151
There are no transcripts associated with this gene.

Gene
PRKCSH
Gene Name
protein kinase C substrate 80K-H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001329.2 Intron NP_001001329.1
NM_001289102.1 Intron NP_001276031.1
NM_001289103.1 Intron NP_001276032.1
NM_001289104.1 Intron NP_001276033.1
NM_002743.3 Intron NP_002734.2
XM_011528130.1 Intron XP_011526432.1
XM_011528131.1 Intron XP_011526433.1
XM_011528132.1 Intron XP_011526434.1
XM_017026977.1 Intron XP_016882466.1

View Full Product Details