Product Details

SNP ID
hCV25612549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:33548579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTCAGGAGGCAGGAAGTGGGG[A/G]CACATCTGGCACTGGTCTTGCACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF122 PubMed Links

Gene Details

Gene
RNF122
Gene Name
ring finger protein 122
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024787.3 1047 UTR 3 NP_079063.2

View Full Product Details