Product Details

SNP ID
rs185404257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39966357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGACGGTGAAGGTGAAGGGAAC[A/G]GCAGGGCTCTCGCAGAACAGCACTC
Phenotype
MIM: 611218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMA PubMed Links

Gene Details

Gene
GSDMA
Gene Name
gasdermin A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178171.4 430 Silent Mutation ACA,ACG T104T NP_835465.2
XM_006721832.2 430 Silent Mutation ACA,ACG T104T XP_006721895.1
XM_011524651.2 430 Intron XP_011522953.1
XM_017024502.1 430 Silent Mutation ACA,ACG T104T XP_016879991.1
XM_017024503.1 430 Silent Mutation ACA,ACG T104T XP_016879992.1
XM_017024504.1 430 Silent Mutation ACA,ACG T104T XP_016879993.1

View Full Product Details