Product Details
- SNP ID
-
rs192323920
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:37486632 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCCCTCCCAGTGACAGTCTAGT[C/G]CCACCGTACATTGTGTGGTACATTG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
POLR1E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149832071] are located under a probe and SNP(s) [rs149832071] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POLR1E
- Gene Name
- RNA polymerase I subunit E
View Full Product Details