Product Details

SNP ID
rs78823293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28348962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACACAACAATATAGCAGATAAG[C/G]CTCCCTCTAAGAATGTAACTTGGCC
Phenotype
MIM: 611519 MIM: 616815 MIM: 191161
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLDIP2 PubMed Links

Gene Details

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290145.1 Intron NP_001277074.1
NM_015584.4 Intron NP_056399.1
Gene
TMEM199
Gene Name
transmembrane protein 199
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details