Product Details

SNP ID
rs34108350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28349157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCAGGCTGCTCCTTGGATAACAC[C/T]GGTTCCTGTAAAGGTTTAGGCAAAA
Phenotype
MIM: 611519 MIM: 616815 MIM: 191161
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLDIP2 PubMed Links

Gene Details

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290145.1 1005 Silent Mutation CCA,CCG P288P NP_001277074.1
NM_015584.4 1005 Silent Mutation CCA,CCG P306P NP_056399.1
Gene
TMEM199
Gene Name
transmembrane protein 199
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details