Product Details

SNP ID
rs115275682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28350926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCTGAGAGTATCTGTGGGATAA[A/G]GGTGATAGCTGAACCATGGCCTACA
Phenotype
MIM: 611519 MIM: 616815 MIM: 191161
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4723 PubMed Links

Gene Details

Gene
MIR4723
Gene Name
microRNA 4723
There are no transcripts associated with this gene.

Gene
POLDIP2
Gene Name
DNA polymerase delta interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290145.1 Intron NP_001277074.1
NM_015584.4 Intron NP_056399.1
Gene
TMEM199
Gene Name
transmembrane protein 199
There are no transcripts associated with this gene.

Gene
TNFAIP1
Gene Name
TNF alpha induced protein 1
There are no transcripts associated with this gene.

View Full Product Details