Product Details
- SNP ID
-
rs62640053
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44316631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCTGCCGCAGACACAGCTTCAT[A/G]AGCACGGAGCCGCTGTCAGCTGAAG
- Phenotype
-
MIM: 605448
MIM: 610820
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RUNDC3A
PubMed Links
Gene Details
- Gene
- RUNDC3A
- Gene Name
- RUN domain containing 3A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144825.1 |
1299 |
Missense Mutation |
ATA,ATG |
I368M |
NP_001138297.1 |
NM_001144826.1 |
1299 |
Missense Mutation |
ATA,ATG |
I363M |
NP_001138298.1 |
NM_006695.4 |
1299 |
Missense Mutation |
ATA,ATG |
I368M |
NP_006686.1 |
XM_005256964.3 |
1299 |
Missense Mutation |
ATA,ATG |
I363M |
XP_005257021.1 |
XM_017024035.1 |
1299 |
Missense Mutation |
ATA,ATG |
I368M |
XP_016879524.1 |
XM_017024036.1 |
1299 |
Missense Mutation |
ATA,ATG |
I364M |
XP_016879525.1 |
XM_017024037.1 |
1299 |
Missense Mutation |
ATA,ATG |
I363M |
XP_016879526.1 |
XM_017024038.1 |
1299 |
Missense Mutation |
ATA,ATG |
I368M |
XP_016879527.1 |
XM_017024039.1 |
1299 |
Missense Mutation |
ATA,ATG |
I368M |
XP_016879528.1 |
XM_017024040.1 |
1299 |
Missense Mutation |
ATA,ATG |
I364M |
XP_016879529.1 |
- Gene
- RUNDC3A-AS1
- Gene Name
- RUNDC3A antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SLC25A39
- Gene Name
- solute carrier family 25 member 39
There are no transcripts associated with this gene.
View Full Product Details