Product Details

SNP ID
rs77229224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114093395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGTCACCCCTCCAGGCTGGGC[C/T]GTGAGACTCACATGGCAAGGGGGAG
Phenotype
MIM: 611278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 Intron NP_612433.1
XM_005251683.4 Intron XP_005251740.1
XM_006716947.2 Intron XP_006717010.1
XM_011518190.2 Intron XP_011516492.1
XM_011518191.2 Intron XP_011516493.1
XM_011518192.2 Intron XP_011516494.1
XM_011518193.2 Intron XP_011516495.1
XM_017014240.1 Intron XP_016869729.1

View Full Product Details