Product Details

SNP ID
rs9619852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22244968 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTTGTGGTCCTCAGCCGGTGCT[A/G]CATCAGCCGCCGGCCATGTCCTCGG
Phenotype
MIM: 605141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPREB1 PubMed Links

Gene Details

Gene
VPREB1
Gene Name
pre-B lymphocyte 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303509.1 89 Silent Mutation CTA,CTG L22L NP_001290438.1
NM_007128.3 89 Silent Mutation CTA,CTG L23L NP_009059.1

View Full Product Details