Product Details

SNP ID
rs34730014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67482702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTAGCAGAAGGCATTGAAGAA[C/G]CGGCAGTAGCACGTGGCACATGGGT
Phenotype
MIM: 602311 MIM: 607028
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGRP PubMed Links

Gene Details

Gene
AGRP
Gene Name
agouti related neuropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001138.1 633 Silent Mutation CGC,CGG R111R NP_001129.1
Gene
ATP6V0D1
Gene Name
ATPase H+ transporting V0 subunit d1
There are no transcripts associated with this gene.

View Full Product Details