Product Details

SNP ID
rs7223058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57867994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACGAGAGACGGGCAGTGGACTGGT[C/G]GCCAAGCTCATGGAGGAGAGGGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CUEDC1 PubMed Links

Gene Details

Gene
CUEDC1
Gene Name
CUE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271875.1 Intron NP_001258804.1
NM_001292025.1 Intron NP_001278954.1
XM_006721900.2 Intron XP_006721963.1
XM_011524812.2 Intron XP_011523114.1
XM_017024640.1 Intron XP_016880129.1
XM_017024641.1 Intron XP_016880130.1

View Full Product Details