Product Details

SNP ID
hCV25614292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108988473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATGGAAAGGGTTTCAGCACCA[C/T]TAGGTTAGGTGAAGGAGACTACGAA
Phenotype
MIM: 606103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC2 PubMed Links

Gene Details

Gene
ARMC2
Gene Name
armadillo repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286609.1 Intron NP_001273538.1
NM_032131.5 Intron NP_115507.4
Gene
SESN1
Gene Name
sestrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199933.1 Intron NP_001186862.1
NM_001199934.1 Intron NP_001186863.1
NM_014454.2 Intron NP_055269.1

View Full Product Details