Product Details

SNP ID
rs34644009
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:52375507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGGCTATGGTGACCAGGCCAGG[G/T]GAGTCAGGCTTCTGCTGGGAGAACT
Phenotype
MIM: 601902
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CC2D1B PubMed Links

Gene Details

Gene
CC2D1B
Gene Name
coiled-coil and C2 domain containing 1B
There are no transcripts associated with this gene.

Gene
ORC1
Gene Name
origin recognition complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190818.1 2349 Silent Mutation TCA,TCC S742S NP_001177747.1
NM_001190819.1 2349 Silent Mutation TCA,TCC S737S NP_001177748.1
NM_004153.3 2349 Silent Mutation TCA,TCC S742S NP_004144.2
XM_011541527.2 2349 Silent Mutation TCA,TCC S382S XP_011539829.1
XM_017001388.1 2349 Silent Mutation TCA,TCC S706S XP_016856877.1
XM_017001389.1 2349 Silent Mutation TCA,TCC S496S XP_016856878.1
Gene
PLA2G12AP1
Gene Name
phospholipase A2 group XIIA pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details