Product Details

SNP ID
rs112956978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88126464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACAGAGAAGGGCACTGTGTCTCC[C/A]AAGGGGCCACAGAAACTCACTGGGG
Phenotype
MIM: 134650
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
FABP1 PubMed Links

Gene Details

Gene
FABP1
Gene Name
fatty acid binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001443.2 Intron NP_001434.1

View Full Product Details