Product Details

SNP ID
rs6732835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85695345 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGGTCTTCTCTCGTCTGAGCCC[C/T]GAGTACTACGACCTGGCAAGAGCCC
Phenotype
MIM: 188855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNLY PubMed Links

Gene Details

Gene
GNLY
Gene Name
granulysin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302758.1 220 Silent Mutation CCC,CCT P53P NP_001289687.1
NM_006433.4 220 Silent Mutation CCC,CCT P26P NP_006424.2
NM_012483.3 220 Silent Mutation CCC,CCT P11P NP_036615.2
XM_005264084.2 220 Silent Mutation CCC,CCT P53P XP_005264141.1
XM_005264085.2 220 Silent Mutation CCC,CCT P26P XP_005264142.1
XM_005264087.2 220 Silent Mutation CCC,CCT P11P XP_005264144.1

View Full Product Details