Product Details

SNP ID
rs61731731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:134918678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCTCCTCTTCACTATCAAAGG[G/T]GACGACACACGTCACTGGACCAAAT
Phenotype
MIM: 606467
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH8A1 PubMed Links

Gene Details

Gene
ALDH8A1
Gene Name
aldehyde dehydrogenase 8 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193480.1 1120 Missense Mutation ACC,CCC T351P NP_001180409.1
NM_022568.3 1120 Missense Mutation ACC,CCC T401P NP_072090.1
NM_170771.2 1120 Missense Mutation ACC,CCC T347P NP_739577.1

View Full Product Details