Product Details

SNP ID
rs61731744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:37570616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTCTTCTCGTGTCTCTTTTTCAG[G/T]TATTTCCACAGGCACTCCATCAATG
Phenotype
MIM: 608777
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POSTN PubMed Links

Gene Details

Gene
POSTN
Gene Name
periostin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135934.1 2346 Missense Mutation ACT,CCT T688P NP_001129406.1
NM_001135935.1 2346 Missense Mutation ACT,CCT T718P NP_001129407.1
NM_001135936.1 2346 Missense Mutation ACT,CCT T688P NP_001129408.1
NM_001286665.1 2346 Missense Mutation ACT,CCT T718P NP_001273594.1
NM_001286666.1 2346 Intron NP_001273595.1
NM_001286667.1 2346 Intron NP_001273596.1
NM_006475.2 2346 Missense Mutation ACT,CCT T745P NP_006466.2
XM_005266231.2 2346 Missense Mutation ACT,CCT T745P XP_005266288.1
XM_005266232.2 2346 Missense Mutation ACT,CCT T715P XP_005266289.1
XM_017020355.1 2346 Missense Mutation ACT,CCT T745P XP_016875844.1
XM_017020356.1 2346 Missense Mutation ACT,CCT T745P XP_016875845.1

View Full Product Details