Product Details

SNP ID
rs200285093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170248875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGGAATGAAAAGCAGGAACTT[C/G]ACTTTCAGTTTTTTTATCTGCATAA
Phenotype
MIM: 601603
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C5orf58 PubMed Links

Gene Details

Gene
C5orf58
Gene Name
chromosome 5 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102609.2 Intron NP_001096079.1
NM_001305393.1 Intron NP_001292322.1
NM_001305394.1 Intron NP_001292323.1
NM_001305395.1 Intron NP_001292324.1
NM_001305396.1 Intron NP_001292325.1
NM_001305397.1 Intron NP_001292326.1
Gene
LCP2
Gene Name
lymphocyte cytosolic protein 2
There are no transcripts associated with this gene.

View Full Product Details