Product Details

SNP ID
rs144906663
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:116393011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCTGAAGATCGATGGGTACAC[A/G]GAGGGCGAGGGCAAGCTGGGGGACA
Phenotype
MIM: 182310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP1A1 PubMed Links

Gene Details

Gene
ATP1A1
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000701.7 Intron NP_000692.2
NM_001160233.1 Intron NP_001153705.1
NM_001160234.1 Intron NP_001153706.1
XM_017001360.1 Intron XP_016856849.1
XM_017001361.1 Intron XP_016856850.1
Gene
ATP1A1-AS1
Gene Name
ATP1A1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details