Product Details

SNP ID
rs55799546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43192227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCCGGCCTAGGCCGGAGGGCGG[A/G]GTTTGCCCTGGGCCGCTGCCGGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NIM1K PubMed Links

Gene Details

Gene
NIM1K
Gene Name
NIM1 serine/threonine protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153361.3 160 UTR 5 NP_699192.1
XM_006714450.1 160 Intron XP_006714513.1
XM_006714451.1 160 UTR 5 XP_006714514.1
XM_017009164.1 160 UTR 5 XP_016864653.1
XM_017009165.1 160 Intron XP_016864654.1

View Full Product Details