Product Details

SNP ID
rs199675608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17874571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGACTCTGGGAGATTAGGGAAG[C/T]ATGTCTGGGAAGAGAAAGGGAGGGA
Phenotype
MIM: 601843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC5A5 PubMed Links

Gene Details

Gene
SLC5A5
Gene Name
solute carrier family 5 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000453.2 Intron NP_000444.1
XM_011528192.2 Intron XP_011526494.1
XM_011528193.2 Intron XP_011526495.1
XM_011528194.2 Intron XP_011526496.1
XM_017027158.1 Intron XP_016882647.1

View Full Product Details