Product Details

SNP ID
rs73223651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:111731123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCATGCTTCTGAAACCATTTTT[G/T]AAAATTTTGCTTTTCCTGCCCAGCC
Phenotype
MIM: 607332
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NREP PubMed Links

Gene Details

Gene
NREP
Gene Name
neuronal regeneration related protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142474.1 Intron NP_001135946.1
NM_001142475.1 Intron NP_001135947.1
NM_001142476.1 Intron NP_001135948.1
NM_001142477.1 Intron NP_001135949.1
NM_001142478.1 Intron NP_001135950.1
NM_001142479.1 Intron NP_001135951.1
NM_001142480.1 Intron NP_001135952.1
NM_001142481.1 Intron NP_001135953.1
NM_001142482.1 Intron NP_001135954.1
NM_001142483.1 Intron NP_001135955.1
NM_004772.2 Intron NP_004763.1
XM_006714732.1 Intron XP_006714795.1
XM_006714733.2 Intron XP_006714796.1
Gene
STARD4-AS1
Gene Name
STARD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details