Product Details
- SNP ID
-
rs7972608
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:51048833 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCTCCAGAGGCATACCCTGAGTT[A/C]CTTTCCTGCCTGGCCTTTGCCTGAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LETMD1
PubMed Links
Gene Details
- Gene
- LETMD1
- Gene Name
- LETM1 domain containing 1
View Full Product Details