Product Details

SNP ID
rs145911686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:107381206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGCAGTTAGGTGGATCTCTGGT[A/G]TTCCAAGACCCTGATGTTTTCTGTG
Phenotype
MIM: 601535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFNA5 PubMed Links

Gene Details

Gene
EFNA5
Gene Name
ephrin A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001962.2 2406 UTR 3 NP_001953.1
XM_006714565.2 2406 UTR 3 XP_006714628.1
XM_011543250.2 2406 UTR 3 XP_011541552.1
XM_011543251.2 2406 UTR 3 XP_011541553.1
XM_017009205.1 2406 UTR 3 XP_016864694.1

View Full Product Details