Product Details

SNP ID
rs964551232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163469491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATGTATCATAGGTAAATATGAT[C/T]ATTCAAATAGCTAGTAACATTAGAA
Phenotype
MIM: 600936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMMR PubMed Links

Gene Details

Gene
HMMR
Gene Name
hyaluronan mediated motility receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142556.1 Intron NP_001136028.1
NM_001142557.1 Intron NP_001136029.1
NM_012484.2 Intron NP_036616.2
NM_012485.2 Intron NP_036617.2
Gene
NUDCD2
Gene Name
NudC domain containing 2
There are no transcripts associated with this gene.

View Full Product Details