Product Details
- SNP ID
-
hCV25618491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:10639451 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTATGACACGACAGTTTAAAGAA[C/T]TACAAGCCCTCAGACTCTACCTAGC
- Phenotype
-
MIM: 601920
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
JAG1
PubMed Links
Gene Details
- Gene
- JAG1
- Gene Name
- jagged 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000214.2 |
4220 |
UTR 3 |
|
|
NP_000205.1 |
View Full Product Details