Product Details

SNP ID
rs144076971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10221595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCTATGTGTAATATGTTTTCA[A/G]TGAAGGATTTTTGTTTGTGAACTCA
Phenotype
MIM: 607420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABARAPL1 PubMed Links

Gene Details

Gene
GABARAPL1
Gene Name
GABA type A receptor associated protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031412.2 Intron NP_113600.1
XM_005253344.4 Intron XP_005253401.1

View Full Product Details