Product Details

SNP ID
hCV25619079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:113768541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGTTCAATGATTATGATCATTA[C/G]CATTTAACACACTATATTATAATGC
Phenotype
MIM: 604950 MIM: 615858
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PHTF1 PubMed Links

Gene Details

Gene
PHTF1
Gene Name
putative homeodomain transcription factor 1
There are no transcripts associated with this gene.

Gene
RSBN1
Gene Name
round spermatid basic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018364.4 Intron NP_060834.2

View Full Product Details