Product Details

SNP ID
rs114671846
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168488843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTATATCCTATGGAATTTGGAAA[A/G]GAAGCATAGAAACCCTATTCTTTTC
Phenotype
MIM: 107820
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RARS PubMed Links

Gene Details

Gene
RARS
Gene Name
arginyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002887.3 Intron NP_002878.2

View Full Product Details