Product Details
- SNP ID
-
hCV25619315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:45094933 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCCCTGCCTGGCGGGCCCTGACA[C/T]ACTAGCATGGTGGTCCTGAGGTCGA
- Phenotype
-
MIM: 606759
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DUOX2
PubMed Links
Gene Details
- Gene
- DUOX2
- Gene Name
- dual oxidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014080.4 |
|
Intron |
|
|
NP_054799.4 |
View Full Product Details