Product Details

SNP ID
rs4938621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119021567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTCTTAATCAGGTGGGTGACATAA[C/T]AAAGTTACTTCACTTCTCTGGTCTC
Phenotype
MIM: 180465 MIM: 602671 MIM: 610971
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
There are no transcripts associated with this gene.

Gene
MIR3656
Gene Name
microRNA 3656
There are no transcripts associated with this gene.

Gene
RPS25
Gene Name
ribosomal protein S25
There are no transcripts associated with this gene.

Gene
SLC37A4
Gene Name
solute carrier family 37 member 4
There are no transcripts associated with this gene.

Gene
TRAPPC4
Gene Name
trafficking protein particle complex 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318486.1 Intron NP_001305415.1
NM_001318488.1 Intron NP_001305417.1
NM_001318489.1 Intron NP_001305418.1
NM_001318490.1 Intron NP_001305419.1
NM_001318492.1 Intron NP_001305421.1
NM_001318494.1 Intron NP_001305423.1
NM_016146.5 Intron NP_057230.1

View Full Product Details