Product Details

SNP ID
rs57880593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:121801541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGCTGCGTGGGCGCTCGGGTCTC[A/C]AGGTGTGGACTGATACCTGGCCCGC
Phenotype
MIM: 606180
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EXOSC9 PubMed Links

Gene Details

Gene
EXOSC9
Gene Name
exosome component 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034194.1 Intron NP_001029366.1
NM_005033.2 Intron NP_005024.2
XM_011532035.2 Intron XP_011530337.1

View Full Product Details