Product Details
- SNP ID
-
hCV25620710
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:37827032 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGGCCGGGAAGGCTTGTCCCCGGA[C/G]GAAACCAGCCATCCGGCCGGCCGGA
- Phenotype
-
MIM: 172000
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PGM2
PubMed Links
Gene Details
- Gene
- PGM2
- Gene Name
- phosphoglucomutase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018290.3 |
|
Intron |
|
|
NP_060760.2 |
View Full Product Details