Product Details

SNP ID
rs61731974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15160815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCCCAGTGGCAGTGGCTGGGCT[A/G]GGCGTGGATTCGGACCAGTCTGAGA
Phenotype
MIM: 600276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOTCH3 PubMed Links

Gene Details

Gene
NOTCH3
Gene Name
notch 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000435.2 6672 Silent Mutation CCC,CCT P2271P NP_000426.2
XM_005259924.4 6672 Silent Mutation CCC,CCT P2219P XP_005259981.1

View Full Product Details