Product Details
- SNP ID
-
rs61731975
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:15160875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGAGGGGCTGGCCCAGTGCTCAGG[A/G]GATTCGGGGGATGGGGTCAGGTAAG
- Phenotype
-
MIM: 600276
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NOTCH3
PubMed Links
Gene Details
- Gene
- NOTCH3
- Gene Name
- notch 3
View Full Product Details